Learn More
Cayman Chemical Aspirin, Purity: ≥90%, CAS: 50-78-2, 500 g
SDP
Catalog No. 502896698
Encompass_Preferred
An NSAID and a COX-1 and COX-2 inhibitor (IC50s = 4.45 and 13.88 µM, respectively, for the human enzymes); an inhibitor of H-PGDS (IC50 = 750 µM for the ovine enzyme); inhibits epinephrine- and ADP-induced platelet aggregation at 6 µg/ml; reduces infarct volume and microglial infiltration in a rat model of ischemia-reperfusion injury induced by MCAO at 30 mg/kg; decreases macrophage infiltration into, increases the number of smooth muscle cells and levels of collagen in, and reduces the area of, atherosclerotic lesions in LDL receptor-deficient mice fed a high-fat diet in the drinking water at 30 mg/L
Catalog No. 50-289-6698 Supplier Cayman Chemical Supplier No. 70260500 G
An NSAID and a COX-1 and COX-2 inhibitor (IC50s = 4.45 and 13.88 µM, respectively, for the human enzymes); an inhibitor of H-PGDS (IC50 = 750 µM for the ovine enzyme); inhibits epinephrine- and ADP-induced platelet aggregation at 6 µg/ml; reduces infarct volume and microglial infiltration in a rat model of ischemia-reperfusion injury induced by MCAO at 30 mg/kg; decreases macrophage infiltration into, increases the number of smooth muscle cells and levels of collagen in, and reduces the area of, atherosclerotic lesions in LDL receptor-deficient mice fed a high-fat diet in the drinking water at 30 mg/L