Learn More
Cayman Chemical SynthetIc Aurapten 10mg
SDP
Catalog No. 502952876
Encompass_Preferred
A coumarin derived from citrus plants with anti-inflammatory, anti-carcinogenic, anti-bacterial, neuroprotective, and hepatoprotective activities; inhibits leukocyte activation and induces phase II enzymes during the initiation phase of carcinogenesis
Catalog No. 50-295-2876 Supplier Cayman Chemical Supplier No. 1400010 MG
A coumarin derived from citrus plants with anti-inflammatory, anti-carcinogenic, anti-bacterial, neuroprotective, and hepatoprotective activities; inhibits leukocyte activation and induces phase II enzymes during the initiation phase of carcinogenesis