Learn More
Cayman Chemical Bergaptol, Purity: ≥95%, CAS: 486-60-2, 10 mg
SDP
Catalog No. 502917162
Encompass_Preferred
A furanocoumarin with diverse biological activities; inhibits CYP3A4 (IC50 = 24.92 µM); scavenges DPPH and ABTS radicals in cell-free assays (EC50s = 26.66 and 13.55 µg/ml, respectively); reduces the replication of L. donovani, T. b. brucei, and T. cruzi in L6 myoblasts (IC50s = 2.5, 22.1, and 68 µg/ml, respectively); induces cell cycle arrest in the S phase and apoptosis in MCF-7 cells in a concentration-dependent manner; decreases the proliferation of HeLa and HepG2 cells (IC50s = 58.57 and 68.42 µM, respectively)
Catalog No. 50-291-7162 Supplier Cayman Chemical Supplier No. 3874210 MG
A furanocoumarin with diverse biological activities; inhibits CYP3A4 (IC50 = 24.92 µM); scavenges DPPH and ABTS radicals in cell-free assays (EC50s = 26.66 and 13.55 µg/ml, respectively); reduces the replication of L. donovani, T. b. brucei, and T. cruzi in L6 myoblasts (IC50s = 2.5, 22.1, and 68 µg/ml, respectively); induces cell cycle arrest in the S phase and apoptosis in MCF-7 cells in a concentration-dependent manner; decreases the proliferation of HeLa and HepG2 cells (IC50s = 58.57 and 68.42 µM, respectively)