Learn More
ATCC Synthetic BK; polyomavirus DNA
Catalog No. 502384606
Encompass
Quantitative synthetic DNA for BK virus can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load and for other molecular biology applications. This preparation includes the full-length genome of BK virus derived from a plasmid clone (ATCC 45025).
Catalog No. 50-238-4606 Supplier ATCC Supplier No. VR3249SD
Quantitative synthetic DNA for BK virus can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load and for other molecular biology applications. This preparation includes the full-length genome of BK virus derived from a plasmid clone (ATCC 45025).