Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Synthetic Block (10X)

Catalog No. PA017
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Bottle, 75 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA017 1 Bottle, 75 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA017 Supplier Invitrogen™ Supplier No. PA017
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Invitrogen's 10X Synthetic Block is a concentrated polymer-based protein-free blocking agent.

Invitrogen's 10X Synthetic Block is a concentrated polymer-based protein-free blocking agent. This easy to use blocking agent has been optimized for use with Invitrogen's ProtoArray Human Protein Microarrays and results in optimal assay performance and signal-to-background ratio. Sufficient reagent for processing 10 arrays.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Packaging Type Bottle
Dilution 10X
Quantity 1 Bottle, 75 mL
Solution Type Blocking/Background Suppression Reagent
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.