Learn More
Mopec Synthetic Blood
SDP
Catalog No. 22446338 Shop All Mopec Products
Change view
Click to view available options
Quantity:
8 oz.
Catalog No. Quantity
22-446-338 8 oz.
1 options
Catalog No. 22-446-338 Supplier Mopec Supplier No. FS356
Add to Cart
Add to Cart
Formulated for classroom forensic training without pathogens from human or non-human blood, this material mimics the chemical composition and appearance of crime scene blood and produces positive reactions with common blood tests and protein enhancement techniques.
Performance
  • Mimics the chemical composition and appearance of crime scene blood
  • Gives positive reactions to common chemical tests for blood
Capacity
  • 8 oz.
Specifications
Quantity 8 oz.
Class Size 1 Lab Group
Product Type Synthetic Blood