Learn More
Cayman Chemical SynthetIc BufotalIn 10mg
SDP
Catalog No. 502946282
Encompass_Preferred
A bufadienolide with cytotoxic and immunomodulatory activities; induces cell cycle arrest at the G1 phase and apoptosis in Hep3B cells from 0.13-1.13 µM; reduces autoantibody production, serum levels of TNF-α, IL-1β, IFN-γ, IL-22, and IL-17, and the number of splenic and lymph node Th17 cells, as well as increases saliva flow rates in a mouse model of experimental Sjörgren’s syndrome at 100 UG/kg; induces lethality in anesthetized dogs (LD50 = 0.36 mg/kg)
Catalog No. 50-294-6282 Supplier Cayman Chemical Supplier No. 3446610 MG
A bufadienolide with cytotoxic and immunomodulatory activities; induces cell cycle arrest at the G1 phase and apoptosis in Hep3B cells from 0.13-1.13 µM; reduces autoantibody production, serum levels of TNF-α, IL-1β, IFN-γ, IL-22, and IL-17, and the number of splenic and lymph node Th17 cells, as well as increases saliva flow rates in a mouse model of experimental Sjörgren’s syndrome at 100 UG/kg; induces lethality in anesthetized dogs (LD50 = 0.36 mg/kg)