Learn More
Cayman Chemical SynthetIc ButIn 1mg
SDP
Catalog No. 502945928
Encompass_Preferred
A flavanone with diverse biological activities; induces melanogenesis in B16/F10 melanoma cells at 1 µM; decreases cytochalasin B-induced DNA damage in isolated human peripheral blood lymphocytes from 1-4 UG/ml; inhibits implantation and decreases the number of pups when administered to pregnant rats from 5-20 mg/animal; increases uterine weight in ovariectomized rats at 0.5 mg/animal
Catalog No. 50-294-5928 Supplier Cayman Chemical Supplier No. 402681 MG
A flavanone with diverse biological activities; induces melanogenesis in B16/F10 melanoma cells at 1 µM; decreases cytochalasin B-induced DNA damage in isolated human peripheral blood lymphocytes from 1-4 UG/ml; inhibits implantation and decreases the number of pups when administered to pregnant rats from 5-20 mg/animal; increases uterine weight in ovariectomized rats at 0.5 mg/animal