Learn More
Cayman Chemical SynthetIc C82 10mg
SDP
Catalog No. 502950245
Encompass_Preferred
An Mtb CdnP inhibitor (IC50 = 17.5 µM); selective for Mtb CdnP over Yybt, RocR, and GBS CdnP, ENPP1, and poxin at 200 µM
Catalog No. 50-295-0245 Supplier Cayman Chemical Supplier No. 3356310 MG
An Mtb CdnP inhibitor (IC50 = 17.5 µM); selective for Mtb CdnP over Yybt, RocR, and GBS CdnP, ENPP1, and poxin at 200 µM