Learn More
Cayman Chemical SynthetIc CalcItrIol 10mg
SDP
Catalog No. 502893917
Encompass_Preferred
An agonist of the vitamin D3 receptor and active metabolite of vitamin D3; binds to the vitamin D3 receptor (Kd = 16 pM) and induces differentiation of U937 cells (EC50 = 2,000 pM); induces cell cycle arrest at the S phase and apoptosis in primary adenomatous and hyperplastic human parathyroid cells at 10 nM; increases bone calcium mobilization in chicks on a low calcium diet at 0.65 nmol/animal; stimulates calcium absorption in the intestines in a chick model of rickets induced by a vitamin D3-deficient diet
Catalog No. 50-289-3917 Supplier Cayman Chemical Supplier No. 7182010 MG
An agonist of the vitamin D3 receptor and active metabolite of vitamin D3; binds to the vitamin D3 receptor (Kd = 16 pM) and induces differentiation of U937 cells (EC50 = 2,000 pM); induces cell cycle arrest at the S phase and apoptosis in primary adenomatous and hyperplastic human parathyroid cells at 10 nM; increases bone calcium mobilization in chicks on a low calcium diet at 0.65 nmol/animal; stimulates calcium absorption in the intestines in a chick model of rickets induced by a vitamin D3-deficient diet