Learn More
Cayman Chemical Carubicin, Purity: ≥98%, CAS: 50935-04-1, 10 mg
SDP
Catalog No. 502938043
Encompass_Preferred
An anthracycline with anticancer activity; cytotoxic to MCF-7 and K562 cells (IC50s = 90 and 60 nM, respectively), as well as P-glycoprotein-expressing MCF-7/DOX and K562i/S9 cells (IC50s = 90 and 60 nM, respectively)
Catalog No. 50-293-8043 Supplier Cayman Chemical Supplier No. 2766010 MG
An anthracycline with anticancer activity; cytotoxic to MCF-7 and K562 cells (IC50s = 90 and 60 nM, respectively), as well as P-glycoprotein-expressing MCF-7/DOX and K562i/S9 cells (IC50s = 90 and 60 nM, respectively)