Learn More
Cayman Chemical SynthetIc CarubIcIn 1mg
SDP
Catalog No. 502940265
Encompass_Preferred
An anthracycline with anticancer activity; cytotoxic to MCF-7 and K562 cells (IC50s = 90 and 60 nM, respectively), as well as P-glycoprotein-expressing MCF-7/DOX and K562i/S9 cells (IC50s = 90 and 60 nM, respectively)
Catalog No. 50-294-0265 Supplier Cayman Chemical Supplier No. 276601 MG
An anthracycline with anticancer activity; cytotoxic to MCF-7 and K562 cells (IC50s = 90 and 60 nM, respectively), as well as P-glycoprotein-expressing MCF-7/DOX and K562i/S9 cells (IC50s = 90 and 60 nM, respectively)