Learn More
Cayman Chemical SynthetIc Catechol 100g
SDP
Catalog No. 502910175
Encompass_Preferred
A polyphenol; has been found in teas, fruits, and vegetables, as well as cigarette smoke, and is used in rubber and plastic production; used as a precursor in the synthesis of organic polymerization inhibitors, flavoring and fragrance agents, and pesticides; an organic pollutant; has been found in industrial wastewater
Catalog No. 50-291-0175 Supplier Cayman Chemical Supplier No. 34791100 G
A polyphenol; has been found in teas, fruits, and vegetables, as well as cigarette smoke, and is used in rubber and plastic production; used as a precursor in the synthesis of organic polymerization inhibitors, flavoring and fragrance agents, and pesticides; an organic pollutant; has been found in industrial wastewater