Learn More
Cayman Chemical SynthetIc CBGB 1mg
SDP
Catalog No. 502940950
Encompass_Preferred
An analytical reference standard that is structurally similar to known phytocannabinoids; intended for research and forensic applications
Catalog No. 50-294-0950 Supplier Cayman Chemical Supplier No. 291091 MG
An analytical reference standard that is structurally similar to known phytocannabinoids; intended for research and forensic applications