Learn More
Cayman Chemical SynthetIc CBGM 1mg
SDP
Catalog No. 502894556
Encompass_Preferred
An analytical reference standard categorized as a phytocannabinoid; intended for research and forensic applications
Catalog No. 50-289-4556 Supplier Cayman Chemical Supplier No. 90038931 MG
An analytical reference standard categorized as a phytocannabinoid; intended for research and forensic applications