Learn More
ATCC Synthetic Chikungunya; virus RNA
Supplier:  ATCC VR3246SD
Encompass
ATCC Genuine Nucleics can be used for assay development, verification, validation, monitoring of day to day test variation, and lot to lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. Preparation includes fragments from the 5’ UTR, nsP1, nsP2, nsP3, nsP4, E2, and E1 genes.
Catalog No. 50-238-2596