Learn More
Cayman Chemical Cirsiliol, Purity: ≥90%, CAS: 34334-69-5, 10 mg
SDP
Catalog No. 502991404
Encompass_Preferred
A flavonoid with diverse biological activities; inhibits 5-LO and 12-LO (IC50s = 0.1 and 1 µM, respectively); inhibits release of SRS-A in passively sensitized isolated guinea pig lung (IC50 = 0.4 µM); induces relaxation of isolated precontracted rat uterus, urinary bladder, proximal aorta, and trachea; inhibits colony formation and migration of B16/F10 murine melanoma cells; enhances radiation-induced inhibition of tumor growth in an H1299 NSCLC mouse xenograft model at 200 µg/kg
Catalog No. 50-299-1404 Supplier Cayman Chemical Supplier No. 3087410 MG
A flavonoid with diverse biological activities; inhibits 5-LO and 12-LO (IC50s = 0.1 and 1 µM, respectively); inhibits release of SRS-A in passively sensitized isolated guinea pig lung (IC50 = 0.4 µM); induces relaxation of isolated precontracted rat uterus, urinary bladder, proximal aorta, and trachea; inhibits colony formation and migration of B16/F10 murine melanoma cells; enhances radiation-induced inhibition of tumor growth in an H1299 NSCLC mouse xenograft model at 200 µg/kg