Learn More
MSC Contec Professional Cloths, Half Fold Dry, Spunlace, 75 Sheets/Bucket
Catalog No. 502819924
Encompass
ContecClean® Cloths are engineered from 100% synthetic fibers, offering disinfectant delivery without quat binding and debris-lifting power like traditional microfiber. These half-fold, dry, perfed roll cloths are supplied in a center pull bucket, suitable for cleanroom applications, delicate tasks, and surface preparation before critical tasks.
  • Engineered from 100% synthetic fibers for consistent disinfectant delivery
  • Offers debris-lifting power and surface contact benefits of traditional microfiber
  • Durable and soft, providing an excellent replacement for re-laundered rags
  • Compatible with quats and other common cleaners and disinfectants
  • Designed for single-use to reduce cross-contamination and support compliance
  • Low lint and alcohol-free for critical applications
  • Includes dispensing container
Catalog No. 50-281-9924 Supplier MSC Supplier No. 11571346
ContecClean® Cloths are engineered from 100% synthetic fibers, offering disinfectant delivery without quat binding and debris-lifting power like traditional microfiber. These half-fold, dry, perfed roll cloths are supplied in a center pull bucket, suitable for cleanroom applications, delicate tasks, and surface preparation before critical tasks.
  • Engineered from 100% synthetic fibers for consistent disinfectant delivery
  • Offers debris-lifting power and surface contact benefits of traditional microfiber
  • Durable and soft, providing an excellent replacement for re-laundered rags
  • Compatible with quats and other common cleaners and disinfectants
  • Designed for single-use to reduce cross-contamination and support compliance
  • Low lint and alcohol-free for critical applications
  • Includes dispensing container