Learn More
Cayman Chemical SynthetIc Colupulon 1mg
SDP
Catalog No. 502946890
Encompass_Preferred
A β-acid with diverse biological activities; activates the PXR in a reporter assay at 1, 3, and 10 µM; dietary administration induces hepatic CYP3A activity in mice at 0.18% w/w; decreases serum glucose levels in non-diabetic mice and increases serum glucose levels in db/db mice
Catalog No. 50-294-6890 Supplier Cayman Chemical Supplier No. 340481 MG
A β-acid with diverse biological activities; activates the PXR in a reporter assay at 1, 3, and 10 µM; dietary administration induces hepatic CYP3A activity in mice at 0.18% w/w; decreases serum glucose levels in non-diabetic mice and increases serum glucose levels in db/db mice