Learn More
Cayman Chemical SynthetIc Curzeren 1mg
SDP
Catalog No. 502911226
Encompass_Preferred
A sesquiterpene with diverse biological activities; scavenges DPPH radicals in a cell-free assay at 3.36 UG/L with a synergistic effect when used in combination with eucalyptol; active against F. graminearum at 0.5 mg/ml; inhibits the proliferation of SK-MEL-19 melanoma cells (IC50 = 5.17 UG/ml), as well as AGP-01 gastric and HCT116 colon cancer cells (IC50s = 8.04 and 9.18 UG/ml, respectively) and MRC-5 non-cancerous human fibroblasts (IC50 = 11.45 UG/ml); induces apoptosis in, and decreases migration of, SK-MEL-19 cells at 5 and 10 µM; reduces tumor weight in an SPC-A1 mouse xenograft model at 135 mg/kg
Catalog No. 50-291-1226 Supplier Cayman Chemical Supplier No. 358281 MG
A sesquiterpene with diverse biological activities; scavenges DPPH radicals in a cell-free assay at 3.36 UG/L with a synergistic effect when used in combination with eucalyptol; active against F. graminearum at 0.5 mg/ml; inhibits the proliferation of SK-MEL-19 melanoma cells (IC50 = 5.17 UG/ml), as well as AGP-01 gastric and HCT116 colon cancer cells (IC50s = 8.04 and 9.18 UG/ml, respectively) and MRC-5 non-cancerous human fibroblasts (IC50 = 11.45 UG/ml); induces apoptosis in, and decreases migration of, SK-MEL-19 cells at 5 and 10 µM; reduces tumor weight in an SPC-A1 mouse xenograft model at 135 mg/kg