Learn More
ATCC Synthetic Cyclospora; cayetanensis DNA
Catalog No. 502383934
Encompass
Quantitative Synthetic DNA from Cyclospora cayetanensis can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine protozoan load and for other molecular biology applications. Preparation includes the full 18S rRNA gene sequence, and full ITS1 and ITS2 sequences.
Catalog No. 50-238-3934 Supplier ATCC Supplier No. PRA3000SD
Quantitative Synthetic DNA from Cyclospora cayetanensis can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine protozoan load and for other molecular biology applications. Preparation includes the full 18S rRNA gene sequence, and full ITS1 and ITS2 sequences.