Learn More
Cayman Chemical SynthetIc DamInozIde 100mg
SDP
Catalog No. 502954914
Encompass_Preferred
An inhibitor of KDM2A, PHF8, and KDM7A (IC50s = 1.5, 0.55, and 2.1 µM, respectively) and a plant growth regulator; selective for KDM2A, PHF8, and KDM7A over KDM3-6 (IC50s = >100 µM for all), as well as FIH, EGLN1, and BBOX1 (IC50s = >100 µM for all); reduces peduncle and internodal elongation in C. officinalis when applied at the visible bud stage at 1,500, 3,500, or 5,000 ppm
Catalog No. 50-295-4914 Supplier Cayman Chemical Supplier No. 12033100 MG
An inhibitor of KDM2A, PHF8, and KDM7A (IC50s = 1.5, 0.55, and 2.1 µM, respectively) and a plant growth regulator; selective for KDM2A, PHF8, and KDM7A over KDM3-6 (IC50s = >100 µM for all), as well as FIH, EGLN1, and BBOX1 (IC50s = >100 µM for all); reduces peduncle and internodal elongation in C. officinalis when applied at the visible bud stage at 1,500, 3,500, or 5,000 ppm