Learn More
ATCC Synthetic Dengue virus; type 3 RNA
Supplier:  ATCC VR3230SD
Encompass
Quantitative synthetic RNA for Dengue virus type 3 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. This preparation includes fragments from the capsid, membrane, and envelope regions.
Catalog No. 50-238-2223