Learn More
Cayman Chemical SynthetIc DIctamnIn 10mg
SDP
Catalog No. 502946779
Encompass_Preferred
An alkaloid with diverse biological activities; inhibits arachidonic acid-, collagen-, thrombin-, or PAF-induced platelet aggregation in washed isolated rabbit platelets at 100 UG/ml; cytotoxic to human KB oral, HeLa cervical, and DLD-1 colon cancer cells (EC50s = 20.55, 2.5, and 6.4 UG/ml, respectively) and induces apoptosis in A549, PC-9, and EBC-1 human lung cancer cells from 50-200 µM; increases serum ALT and ALP activities and induces hepatocyte necrosis in mice at 150 or 200 mg/kg
Catalog No. 50-294-6779 Supplier Cayman Chemical Supplier No. 3464610 MG
An alkaloid with diverse biological activities; inhibits arachidonic acid-, collagen-, thrombin-, or PAF-induced platelet aggregation in washed isolated rabbit platelets at 100 UG/ml; cytotoxic to human KB oral, HeLa cervical, and DLD-1 colon cancer cells (EC50s = 20.55, 2.5, and 6.4 UG/ml, respectively) and induces apoptosis in A549, PC-9, and EBC-1 human lung cancer cells from 50-200 µM; increases serum ALT and ALP activities and induces hepatocyte necrosis in mice at 150 or 200 mg/kg