Learn More
Cayman Chemical SynthetIc DIMBOA 10mg
SDP
Catalog No. 502918264
Encompass_Preferred
A benzoxazinoid with antimicrobial and antioxidant activities; active against S. aureus, E. faecalis, E. coli, K. pneumoniae, B. subtilis, and P. aeruginosa, as well as S. cerevisiae, in a disc assay at 2 mg/disc; scavenges DPPH radicals in a cell-free assay (IC50 = 11.31 UG/ml)
Catalog No. 50-291-8264 Supplier Cayman Chemical Supplier No. 3964710 MG
A benzoxazinoid with antimicrobial and antioxidant activities; active against S. aureus, E. faecalis, E. coli, K. pneumoniae, B. subtilis, and P. aeruginosa, as well as S. cerevisiae, in a disc assay at 2 mg/disc; scavenges DPPH radicals in a cell-free assay (IC50 = 11.31 UG/ml)