Learn More
SPECTRUM QUALITY STANDARDS Synthetic STD, 1 L
Catalog No. NC1063480
Encompass_Preferred
The automatic distillation apparatus duplicates the distillation conditions of the manual method. The increased reliance on electronic control requires an independent standard to verify the apparatus is performing correctly. Spectrum has introduced a synthetic blend of Hydrocarbons that boil in the temperature range specified in ASTM D86 Distillation Groups 1 and 2 and ASTM D850.
Catalog No. NC1063480 Supplier SPECTRUM QUALITY STANDARDS Supplier No. D862
The automatic distillation apparatus duplicates the distillation conditions of the manual method. The increased reliance on electronic control requires an independent standard to verify the apparatus is performing correctly. Spectrum has introduced a synthetic blend of Hydrocarbons that boil in the temperature range specified in ASTM D86 Distillation Groups 1 and 2 and ASTM D850.