Learn More
Cayman Chemical SynthetIc ErysolIn 10mg
SDP
Catalog No. 502917249
Encompass_Preferred
An isothiocyanate with anticancer activity; inhibits the proliferation of MCF-7, HeLa, A549, HepG2, and SW480 cells (IC50s = 1.62, 1.12, 5.22, 14, and 8.14 µM, respectively); enhances arsenic trioxide-induced apoptosis and cytotoxicity in HL-60, U937, and K562 cells at 3 µM
Catalog No. 50-291-7249 Supplier Cayman Chemical Supplier No. 3966110 MG
An isothiocyanate with anticancer activity; inhibits the proliferation of MCF-7, HeLa, A549, HepG2, and SW480 cells (IC50s = 1.62, 1.12, 5.22, 14, and 8.14 µM, respectively); enhances arsenic trioxide-induced apoptosis and cytotoxicity in HL-60, U937, and K562 cells at 3 µM