Learn More
Cayman Chemical Fisetin-d5, Purity: ≥99% deuterated forms (d1-d5), CAS: 2909407-29-8, 5 mg
SDP
Catalog No. 502910259
Encompass_Preferred
An internal standard for the quantification of fisetin by GC- or LC-MS
Catalog No. 50-291-0259 Supplier Cayman Chemical Supplier No. 347065 MG
An internal standard for the quantification of fisetin by GC- or LC-MS