Learn More
Cayman Chemical SynthetIc FustIn 10mg
SDP
Catalog No. 502912850
Encompass_Preferred
A flavanone with diverse biological activities; enhances epigallocatechin-3-O-gallate-induced decreases in the viability of U266 cells at 5 and 10 µM; inhibits increases in blood glucose levels, as well as serum total cholesterol, triglyceride, and MDA levels, in a mouse model of streptozotocin-induced diabetes at 50 and 100 mg/kg; prevents Aβ42-induced decreases in contextual- and tone fear conditioning freezing time in mice at 100 mg/kg
Catalog No. 50-291-2850 Supplier Cayman Chemical Supplier No. 3627310 MG
A flavanone with diverse biological activities; enhances epigallocatechin-3-O-gallate-induced decreases in the viability of U266 cells at 5 and 10 µM; inhibits increases in blood glucose levels, as well as serum total cholesterol, triglyceride, and MDA levels, in a mouse model of streptozotocin-induced diabetes at 50 and 100 mg/kg; prevents Aβ42-induced decreases in contextual- and tone fear conditioning freezing time in mice at 100 mg/kg