Learn More
Cayman Chemical Fustin, Purity: ≥98%, CAS: 20725-03-5, 5 mg
SDP
Catalog No. 502911696
Encompass_Preferred
A flavanone with diverse biological activities; enhances epigallocatechin-3-O-gallate-induced decreases in the viability of U266 cells at 5 and 10 µM; inhibits increases in blood glucose levels, as well as serum total cholesterol, triglyceride, and MDA levels, in a mouse model of streptozotocin-induced diabetes at 50 and 100 mg/kg; prevents Aβ42-induced decreases in contextual- and tone fear conditioning freezing time in mice at 100 mg/kg
Catalog No. 50-291-1696 Supplier Cayman Chemical Supplier No. 362735 MG
A flavanone with diverse biological activities; enhances epigallocatechin-3-O-gallate-induced decreases in the viability of U266 cells at 5 and 10 µM; inhibits increases in blood glucose levels, as well as serum total cholesterol, triglyceride, and MDA levels, in a mouse model of streptozotocin-induced diabetes at 50 and 100 mg/kg; prevents Aβ42-induced decreases in contextual- and tone fear conditioning freezing time in mice at 100 mg/kg