Learn More
Cayman Chemical Geniposide, Purity: ≥98%, CAS: 24512-63-8, 500 mg
SDP
Catalog No. 502941548
Encompass_Preferred
An iridoid glycoside with diverse biological activities; reduces exudate volume and nitrite levels in a rat model of carrageenan-induced air pouch inflammation at 0.1 mg/pouch; prevents prevents carrageenan-induced paw edema in rats at 100 mg/kg, p.o.; reduces serum levels of ALT and AST and hepatic levels of MDA as well as decreases in hepatic levels of SOD and GPX in a rat model of high-fat diet-induced NASH at 50 and 100 mg/kg; prevents increases in apoptosis and decreases in the number of dopaminergic neurons in the substantia nigra in a mouse model of MPTP-induced Parkinson's disease at 100 mg/kg; improves motor coordination in the rotarod and swim tests in a rat model of Parkinson's disease
Catalog No. 50-294-1548 Supplier Cayman Chemical Supplier No. 28281500 MG
An iridoid glycoside with diverse biological activities; reduces exudate volume and nitrite levels in a rat model of carrageenan-induced air pouch inflammation at 0.1 mg/pouch; prevents prevents carrageenan-induced paw edema in rats at 100 mg/kg, p.o.; reduces serum levels of ALT and AST and hepatic levels of MDA as well as decreases in hepatic levels of SOD and GPX in a rat model of high-fat diet-induced NASH at 50 and 100 mg/kg; prevents increases in apoptosis and decreases in the number of dopaminergic neurons in the substantia nigra in a mouse model of MPTP-induced Parkinson's disease at 100 mg/kg; improves motor coordination in the rotarod and swim tests in a rat model of Parkinson's disease