Learn More
Cayman Chemical SynthetIc GItoxIn 1mg
SDP
Catalog No. 502918319
Encompass_Preferred
A cardiac glycoside with cardiotonic and antiarrhythmic activities; inhibits Na+/K+-ATPase in an isoform-specific biphasic manner with IC50 values of 0.298 and 410 µM for the high- and low affinity human erythrocyte isoenzymes, respectively, and 1.18 and 28.5 µM for the high- and low affinity porcine cerebral cortex isoenzymes, respectively; increases contractility and rhythmicity in isolated guinea pig heart in a concentration-dependent manner; a degradation product of digoxin
Catalog No. 50-291-8319 Supplier Cayman Chemical Supplier No. 388881 MG
A cardiac glycoside with cardiotonic and antiarrhythmic activities; inhibits Na+/K+-ATPase in an isoform-specific biphasic manner with IC50 values of 0.298 and 410 µM for the high- and low affinity human erythrocyte isoenzymes, respectively, and 1.18 and 28.5 µM for the high- and low affinity porcine cerebral cortex isoenzymes, respectively; increases contractility and rhythmicity in isolated guinea pig heart in a concentration-dependent manner; a degradation product of digoxin