Learn More
Cayman Chemical SynthetIc GLN-1062 100mg
SDP
Catalog No. 502942618
Encompass_Preferred
A neuroprotective agent and prodrug form of galantamine; active against M. tuberculosis and M. aurum (MIC = 31.25 UG/ml for both); decreases the frequency of scopolamine-induced spontaneous alternations in the T maze continuous alternation task in mice (ED50 = 0.34 mg/kg); inhibits increases in hippocampal and entorhinal cortex Aβ plaque densities in the 5XFAD transgenic mouse model of Alzheimer's disease when administered intranasally at 6 mg/kg per day
Catalog No. 50-294-2618 Supplier Cayman Chemical Supplier No. 41855100 MG
A neuroprotective agent and prodrug form of galantamine; active against M. tuberculosis and M. aurum (MIC = 31.25 UG/ml for both); decreases the frequency of scopolamine-induced spontaneous alternations in the T maze continuous alternation task in mice (ED50 = 0.34 mg/kg); inhibits increases in hippocampal and entorhinal cortex Aβ plaque densities in the 5XFAD transgenic mouse model of Alzheimer's disease when administered intranasally at 6 mg/kg per day