Learn More
Cayman Chemical SynthetIc GosypetIn 1mg
SDP
Catalog No. 502948723
Encompass_Preferred
A flavonoid with diverse biological activities; inhibits MKK3 and MKK6, as well as induces apoptosis and cell cycle arrest at the G2 phase in KYSE-450 and KYSE-510 esophageal cancer cells at 20 µM; scavenges DPPH radicals and inhibits copper-induced lipid peroxidation in cell-free assays; reduces high-fat diet-induced increases in serum cholesterol, triglycerides, and LDL-cholesterol levels, as well as decreases aortic extracellular lipid and foam cell deposits in a rabbit model of atherosclerosis at 10 mg/kg
Catalog No. 50-294-8723 Supplier Cayman Chemical Supplier No. 338401 MG
A flavonoid with diverse biological activities; inhibits MKK3 and MKK6, as well as induces apoptosis and cell cycle arrest at the G2 phase in KYSE-450 and KYSE-510 esophageal cancer cells at 20 µM; scavenges DPPH radicals and inhibits copper-induced lipid peroxidation in cell-free assays; reduces high-fat diet-induced increases in serum cholesterol, triglycerides, and LDL-cholesterol levels, as well as decreases aortic extracellular lipid and foam cell deposits in a rabbit model of atherosclerosis at 10 mg/kg