Learn More
Cayman Chemical SynthetIc GW 9662 1mg
SDP
Catalog No. 502896708
Encompass_Preferred
The peroxisome proliferator-activated receptor γ (PPARγ) is the nuclear receptor responsible for transducing the therapeutic activity of the thiazolidinediones. Thiazolidinediones are a group of structurally related synthetic PPARγ agonists with antidiabetic actions in vivo. Rosiglitazone (BRL 49653) is a prototypical thiazolidinedione and has served as a reference compound for this class. There are many PPARγ agonists, including 15-deoxy-.DELTA.12,14-prostaglandin J2 and azelaoyl PAF, which are naturally derived. However, only a few antagonists have been reported. GW 9662 blocks the PPARγ-induced differentiation of monocytes to osteoclasts by >90% at a dose of 0.1 µM. It is therefore a much more potent antagonist than BADGE, which is another reported PPARγ antagonist.
Catalog No. 50-289-6708 Supplier Cayman Chemical Supplier No. 707851 MG
The peroxisome proliferator-activated receptor γ (PPARγ) is the nuclear receptor responsible for transducing the therapeutic activity of the thiazolidinediones. Thiazolidinediones are a group of structurally related synthetic PPARγ agonists with antidiabetic actions in vivo. Rosiglitazone (BRL 49653) is a prototypical thiazolidinedione and has served as a reference compound for this class. There are many PPARγ agonists, including 15-deoxy-.DELTA.12,14-prostaglandin J2 and azelaoyl PAF, which are naturally derived. However, only a few antagonists have been reported. GW 9662 blocks the PPARγ-induced differentiation of monocytes to osteoclasts by >90% at a dose of 0.1 µM. It is therefore a much more potent antagonist than BADGE, which is another reported PPARγ antagonist.