Learn More
Cayman Chemical SynthetIc GW 9662 50mg
SDP
Catalog No. 502896918
Encompass_Preferred
This assay kit provides the convenience of an all-inclusive cell-based reporter assay system for M-CSFR signaling. In addition to Reporter Cells included are an optimized culture medium for use in reviving the cryopreserved cells a culture medium for use in preparing test sample treatments the physiological activator M-CSF as a positive control Luciferase Detection Reagents and a cell culture-ready assay plate
Catalog No. 50-289-6918 Supplier Cayman Chemical Supplier No. 7078550 MG
This assay kit provides the convenience of an all-inclusive cell-based reporter assay system for M-CSFR signaling. In addition to Reporter Cells included are an optimized culture medium for use in reviving the cryopreserved cells a culture medium for use in preparing test sample treatments the physiological activator M-CSF as a positive control Luciferase Detection Reagents and a cell culture-ready assay plate