Learn More
ATCC Synthetic Hepatitis C; virus RNA
Supplier:  ATCC VR3233SD
Encompass
Quantitative synthetic RNA for Hepatitis C virus can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. This preparation includes fragments from 5’UTR and X-tail region (3’UTR).
Catalog No. 50-238-4672