Learn More
Cayman Chemical SynthetIc HIspIdol 1mg
SDP
Catalog No. 502914982
Encompass_Preferred
An aurone with diverse biological activities; is an inhibitor of tyrosinase, MAO-A, and MAO-B (IC50s = 38.4, 0.26, and 2.45 µM); increases catalase and SOD activity in cell-free assays, as well as decreases intracellular ROS in, and increases the lifespan of, C. elegans at 50 µM; decreases immobility time in forced swim and tail suspension tests in mice at 2, 5, and 15 mg/kg
Catalog No. 50-291-4982 Supplier Cayman Chemical Supplier No. 373731 MG
An aurone with diverse biological activities; is an inhibitor of tyrosinase, MAO-A, and MAO-B (IC50s = 38.4, 0.26, and 2.45 µM); increases catalase and SOD activity in cell-free assays, as well as decreases intracellular ROS in, and increases the lifespan of, C. elegans at 50 µM; decreases immobility time in forced swim and tail suspension tests in mice at 2, 5, and 15 mg/kg