Learn More
Cayman Chemical Hispolon, Purity: ≥95%, CAS: 173933-40-9, 10 mg
SDP
Catalog No. 502916787
Encompass_Preferred
A phenol with diverse biological activities; scavenges superoxide and hydroxyl radicals in cell-free assays, as well as reduces LPS-induced production of ROS in RAW 264.7 cells at 5 and 10 µM; inhibits cell proliferation and induces apoptosis in C6 rat glioma and DBTRG-05MG human glioblastoma cells from 60-80 µM; reduces acetic acid-induced writhing in mice at 20 mg/kg
Catalog No. 50-291-6787 Supplier Cayman Chemical Supplier No. 3841510 MG
A phenol with diverse biological activities; scavenges superoxide and hydroxyl radicals in cell-free assays, as well as reduces LPS-induced production of ROS in RAW 264.7 cells at 5 and 10 µM; inhibits cell proliferation and induces apoptosis in C6 rat glioma and DBTRG-05MG human glioblastoma cells from 60-80 µM; reduces acetic acid-induced writhing in mice at 20 mg/kg