Learn More
ATCC Synthetic Human; bocavirus DNA
Catalog No. 502382199
Encompass
Quantitative synthetic DNA for Human bocavirus can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load and for other molecular biology applications. This preparation includes fragments from the 5’UTR, NS1, NP1, VP1, VP2, and 3’ UTR genes.
Catalog No. 50-238-2199 Supplier ATCC Supplier No. VR3251SD
Quantitative synthetic DNA for Human bocavirus can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load and for other molecular biology applications. This preparation includes fragments from the 5’UTR, NS1, NP1, VP1, VP2, and 3’ UTR genes.