Learn More
ATCC Synthetic Human; papillomavirus 18 DNA
Catalog No. 502383964
Encompass
Quantitative synthetic DNA for Human papillomavirus 18 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. This preparation includes the full-length genome of human papillomavirus type 18 derived from a plasmid clone (ATCC 45152D).
Catalog No. 50-238-3964 Supplier ATCC Supplier No. VR3241SD
Quantitative synthetic DNA for Human papillomavirus 18 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. This preparation includes the full-length genome of human papillomavirus type 18 derived from a plasmid clone (ATCC 45152D).