Learn More
ATCC Synthetic Human; papillomavirus 31 DNA
Supplier:  ATCC VR3256SD
Encompass
Quantitative synthetic Human papillomavirus 31 DNA can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. This preparation includes a full-length genome of human papillomavirus type 31 (HPV-31) derived from a plasmid clone (ATCC 65446 ).
Catalog No. 50-238-3028
 Restricted Item