Learn More
Cayman Chemical (+)-Kavain, Purity: ≥98%, CAS: 500-64-1, 10 mg
SDP
Catalog No. 502934542
Encompass_Preferred
A kavalactone with diverse biological activities; a positive allosteric modulator of α4β2δ subunit-containing GABAA receptors (EC50s = 25 and 59 µM in the presence and absence of 300 µM (+)-kavain); reduces growth of WPMY-1 prostate cancer cells (IC50= 5.2 µg/ml); reduces LPS-induced TNF-α production in primary murine macrophages; reduces hind paw swelling and TNF-α production in a mouse model of collagen-induced arthritis; reduces acetic acid-induced abdominal constrictions in mice (ED50= 15.71 mg/kg)
Catalog No. 50-293-4542 Supplier Cayman Chemical Supplier No. 2652410 MG
A kavalactone with diverse biological activities; a positive allosteric modulator of α4β2δ subunit-containing GABAA receptors (EC50s = 25 and 59 µM in the presence and absence of 300 µM (+)-kavain); reduces growth of WPMY-1 prostate cancer cells (IC50= 5.2 µg/ml); reduces LPS-induced TNF-α production in primary murine macrophages; reduces hind paw swelling and TNF-α production in a mouse model of collagen-induced arthritis; reduces acetic acid-induced abdominal constrictions in mice (ED50= 15.71 mg/kg)