Learn More
Cayman Chemical Khasianine, Purity: ≥98%, CAS: 32449-98-2, 10 mg
SDP
Catalog No. 502917429
Encompass_Preferred
A steroidal glycoside with diverse biological activities; reduces the proliferation of LNCaP and T47D cancer cells (IC50s = 32.1 and 27.5 µM, respectively); inhibits TNF-α-induced nuclear translocation of NF-κB in HaCaT keratinocytes at 50 nM; decreases AST serum levels in a mouse model of carbon tetrachloride-induced liver injury at 0.1 mg/kg; topical administration reduces ear epidermal thickness and scaling, as well as leukocyte and macrophage infiltration, in an imiquimod-induced mouse model of psoriasis at 5 mg/kg
Catalog No. 50-291-7429 Supplier Cayman Chemical Supplier No. 3876210 MG
A steroidal glycoside with diverse biological activities; reduces the proliferation of LNCaP and T47D cancer cells (IC50s = 32.1 and 27.5 µM, respectively); inhibits TNF-α-induced nuclear translocation of NF-κB in HaCaT keratinocytes at 50 nM; decreases AST serum levels in a mouse model of carbon tetrachloride-induced liver injury at 0.1 mg/kg; topical administration reduces ear epidermal thickness and scaling, as well as leukocyte and macrophage infiltration, in an imiquimod-induced mouse model of psoriasis at 5 mg/kg