Learn More
Cayman Chemical SynthetIc Ku-0060648 5mg
SDP
Catalog No. 502971868
Encompass_Preferred
A dual inhibitor of DNA-PK and PI3K
Catalog No. 50-297-1868 Supplier Cayman Chemical Supplier No. 153445 MG
A dual inhibitor of DNA-PK and PI3K