Learn More
Cayman Chemical L-NNA, Purity: ≥99%, CAS: 2149-70-4, 50 g
SDP
Catalog No. 502896784
Encompass_Preferred
L-NNA is an inhibitor of NOS with some selectivity for the neuronal and endothelial isoforms. It exhibits Ki values of 15 nM, 39 nM, and 4.4 µM for nNOS (bovine), eNOS (human), and iNOS (mouse), respectively.{5433,1857,5648}
Catalog No. 50-289-6784 Supplier Cayman Chemical Supplier No. 8022050 G
L-NNA is an inhibitor of NOS with some selectivity for the neuronal and endothelial isoforms. It exhibits Ki values of 15 nM, 39 nM, and 4.4 µM for nNOS (bovine), eNOS (human), and iNOS (mouse), respectively.{5433,1857,5648}