Learn More
Cayman Chemical Lupulone, Purity: ≥95%, CAS: 468-28-0, 10 mg
SDP
Catalog No. 502939372
Encompass_Preferred
A beta-acid; active against B. subtilis and S. aureus (MICs = 1 and 1.2 µg/ml, respectively), and T. b. brucei and L. m. mexicana (IC50s = 0.9 and 4.7 µg/ml, respectively); scavenges DPPH radicals in a cell-free assay and inhibits lipid peroxidation in rat brain homogenates (IC50s = 25 and 39 µM, respectively); reduces proliferation, cell migration, and capillary tube formation in HUVECs at 2.5 to 50 µg/ml; lupulone (40 µg/ml) activates the extrinsic apoptotic death pathway in SW480 and SW620 colon cancer cells.
Catalog No. 50-293-9372 Supplier Cayman Chemical Supplier No. 2848110 MG
A beta-acid; active against B. subtilis and S. aureus (MICs = 1 and 1.2 µg/ml, respectively), and T. b. brucei and L. m. mexicana (IC50s = 0.9 and 4.7 µg/ml, respectively); scavenges DPPH radicals in a cell-free assay and inhibits lipid peroxidation in rat brain homogenates (IC50s = 25 and 39 µM, respectively); reduces proliferation, cell migration, and capillary tube formation in HUVECs at 2.5 to 50 µg/ml; lupulone (40 µg/ml) activates the extrinsic apoptotic death pathway in SW480 and SW620 colon cancer cells.