Learn More
Cayman Chemical MCI-186, Purity: ≥98%, CAS: 89-25-8, 500 g
SDP
Catalog No. 502954754
Encompass_Preferred
A free radical scavenger with diverse protective effects in vivo; reduces damage due to ischemia-reperfusion injury in lung, liver, and brain in animal models of transplant, infection, traumatic brain injury, and stroke
Catalog No. 50-295-4754 Supplier Cayman Chemical Supplier No. 13320500 G
A free radical scavenger with diverse protective effects in vivo; reduces damage due to ischemia-reperfusion injury in lung, liver, and brain in animal models of transplant, infection, traumatic brain injury, and stroke