Learn More
ATCC Synthetic Middle East; respiratory syndrome; coronavirus RNA
Catalog No. 502382774
Encompass
Quantitative synthetic RNA for Middle East respiratory syndrome coronavirus can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load and for other molecular biology applications. This preparation includes fragments from the ORF1ab, ORF5, upper envelope (upE), ORF8b, nucleocapsid (N) protein gene, and 3' UTR regions.
Catalog No. 50-238-2774 Supplier ATCC Supplier No. VR3248SD
Quantitative synthetic RNA for Middle East respiratory syndrome coronavirus can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load and for other molecular biology applications. This preparation includes fragments from the ORF1ab, ORF5, upper envelope (upE), ORF8b, nucleocapsid (N) protein gene, and 3' UTR regions.