Learn More
Cayman Chemical SynthetIc MIltIron 10mg
SDP
Catalog No. 502913592
Encompass_Preferred
A tanshinone; protects lard from thermal oxidation at 0.02%; a partial agonist of the benzodiazepine receptor (IC50 = 0.3 µM); inhibits CES2 (Ki = 0.04 µM) as well as SARS-CoV Mpro and PLpro (IC50s = 21.1 and 30 µM, respectively); cytotoxic to HCT116 colon cancer cells (IC50 = 4.27 µM) and induces mitochondrial dysfunction and ROS production in the same cells; increases the number of punished crossings in the four-plate test, indicating tranquilizing activity, in mice at 10-60 mg/kg
Catalog No. 50-291-3592 Supplier Cayman Chemical Supplier No. 3666310 MG
A tanshinone; protects lard from thermal oxidation at 0.02%; a partial agonist of the benzodiazepine receptor (IC50 = 0.3 µM); inhibits CES2 (Ki = 0.04 µM) as well as SARS-CoV Mpro and PLpro (IC50s = 21.1 and 30 µM, respectively); cytotoxic to HCT116 colon cancer cells (IC50 = 4.27 µM) and induces mitochondrial dysfunction and ROS production in the same cells; increases the number of punished crossings in the four-plate test, indicating tranquilizing activity, in mice at 10-60 mg/kg